Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044703.2 | NP_001038168.1 | 2 - 3 | 822 | 72 | |
NM_001044704.2 | NP_001038169.1 | 3 - 4 | 888 | 72 | ||
NM_001044705.2 | NP_001038170.1 | 2 - 3 | 803 | 72 | ||
NM_011757.3 | NP_035887.2 | 3 - 4 | 907 | 72 | ||
GenBank mRNA | AK146776.1 | - | 3 - 4 | 881 | 72 | |
AK146875.1 | - | 3 - 4 | 868 | 72 | ||
BC010591.1 | - | 2 - 3 | 750 | 3981599 | 72 | |
BU513148.1 | - | 2 - 3 | 764 | 72 | ||
D10630.1 | - | 2 - 3 | 741 | 72 | ||
X63747.1 | - | 2 - 3 | 1598 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |