Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289477.1 | NP_001276406.1 | 7 - 8 | 935 | 67 | |
NM_001289478.1 | NP_001276407.1 | 7 - 8 | 1122 | 67 | ||
NM_001289479.1 | NP_001276408.1 | 8 - 9 | 1233 | 67 | ||
NM_011863.2 | NP_035993.1 | 6 - 7 | 945 | 67 | ||
GenBank mRNA | AK146507.1 | - | 6 - 7 | 818 | 67 | |
AK153296.1 | - | 6 - 7 | 815 | 67 | ||
AK160875.1 | - | 8 - 9 | 1137 | 67 | ||
AK168982.1 | - | 6 - 7 | 818 | 67 | ||
AK169117.1 | - | 7 - 8 | 997 | 67 | ||
AK169124.1 | - | 6 - 7 | 812 | 67 | ||
AK169204.1 | - | 7 - 8 | 997 | 67 | ||
BC066055.1 | - | 7 - 8 | 1001 | 6833756 | 67 | |
U34883.1 | - | 6 - 7 | 822 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |