Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011867.3 | NP_035997.1 | 5 - 6 | 814 | 66 | |
GenBank mRNA | AF167411.1 | - | 5 - 6 | 815 | 66 | |
BC140995.1 | - | 4 - 5 | 662 | 9055607 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |