Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038018.4 | NP_001033107.1 | 7 - 8 | 778 | 74 | |
NM_001112711.2 | NP_001106182.1 | 7 - 8 | 778 | 74 | ||
NM_001286063.1 | NP_001272992.1 | 7 - 8 | 758 | 74 | ||
NM_001286065.1 | NP_001272994.1 | 7 - 8 | 778 | 74 | ||
NM_001286066.1 | NP_001272995.1 | 7 - 8 | 778 | 74 | ||
NM_011938.4 | NP_036068.2 | 7 - 8 | 778 | 74 | ||
GenBank mRNA | AF040747.1 | - | 7 - 8 | 648 | 74 | |
AF040748.1 | - | 7 - 8 | 648 | 74 | ||
AF040749.1 | - | 7 - 8 | 648 | 74 | ||
AK051405.1 | - | 7 - 8 | 760 | 74 | ||
AK146119.1 | - | 7 - 8 | 653 | 74 | ||
BC075719.1 | - | 7 - 8 | 659 | 5031455 | 74 | |
Y15797.1 | - | 7 - 8 | 600 | 74 | ||
Y15798.1 | - | 7 - 8 | 622 | 74 | ||
Y15799.1 | - | 7 - 8 | 605 | 74 | ||
Y15800.1 | - | 7 - 8 | 600 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |