Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011950.2 | NP_036080.2 | 5 - 6 | 544 | 61 | |
GenBank mRNA | AK146993.1 | - | 5 - 6 | 532 | 61 | |
BC001992.1 | - | 5 - 6 | 513 | 3492903 | 61 | |
U81823.1 | - | 5 - 6 | 501 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |