Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF139923.1 | - | 7 - 8 | 1938 | 80 | |
AK135153.1 | - | 7 - 8 | 2127 | 80 | ||
AK137418.1 | - | 8 - 9 | 2263 | 80 | ||
BC117995.1 | - | 7 - 8 | 1938 | 40096022 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |