Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013485.1 | NP_038513.1 | 6 - 7 | 929 | 132 | |
GenBank mRNA | BC011137.1 | - | 6 - 7 | 929 | 4195787 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |