Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013518.4 | NP_038546.2 | 1 - 2 | 649 | 75 | |
GenBank mRNA | AK158782.1 | - | 1 - 2 | 650 | 75 | |
BC099872.1 | - | 1 - 2 | 400 | 40040091 | 75 | |
BC099873.1 | - | 1 - 2 | 400 | 40040090 | 75 | |
BC099874.1 | - | 1 - 2 | 400 | 40040089 | 75 | |
BC099875.1 | - | 1 - 2 | 400 | 40040088 | 75 | |
BC125237.1 | - | 1 - 2 | 400 | 40040117 | 75 | |
D38258.1 | - | 1 - 2 | 361 | 75 | ||
HM988990.1 | - | 1 - 2 | 279 | 75 | ||
S82023.1 | - | 1 - 2 | 334 | 75 | ||
U33535.1 | - | 1 - 2 | 417 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |