Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143669.1 | - | 3 - 4 | 480 | 94 | |
AK153873.1 | - | 3 - 4 | 494 | 94 | ||
BC014720.1 | - | 3 - 4 | 474 | 4924951 | 94 | |
D13565.1 | - | 3 - 4 | 472 | 94 | ||
D13821.1 | - | 3 - 4 | 467 | 94 | ||
KC815469.1 | - | 3 - 4 | 470 | 94 | ||
L20048.1 | - | 3 - 4 | 484 | 94 | ||
X75337.1 | - | 3 - 4 | 497 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |