Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161775.1 | NP_001155247.1 | 38 - 39 | 5655 | 66 | |
NM_013607.2 | NP_038635.2 | 38 - 39 | 5655 | 66 | ||
GenBank mRNA | AK173045.1 | - | 38 - 39 | 5619 | 66 | |
BC026142.1 | - | 38 - 39 | 5640 | 3989495 | 66 | |
D85923.1 | - | 38 - 39 | 5610 | 66 | ||
D85924.1 | - | 38 - 39 | 5610 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |