Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013657.5 | NP_038685.3 | 12 - 13 | 1539 | 85 | |
GenBank mRNA | AK034239.1 | - | 12 - 13 | 1598 | 85 | |
AK077231.1 | - | 1 - 2 | 662 | 85 | ||
AK140348.1 | - | 2075 | 85 | |||
BC066852.1 | - | 12 - 13 | 1539 | 30093230 | 85 | |
X85994.1 | - | 12 - 13 | 1491 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |