Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013803.2 | NP_038831.2 | 3 - 4 | 1021 | 70 | |
GenBank mRNA | AF110178.1 | - | 3 - 4 | 1022 | 70 | |
AF110179.1 | - | 3 - 4 | 1022 | 70 | ||
AF128842.1 | - | 2 - 3 | 719 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mus musculus Probe spans exons |