Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013820.3 | NP_038848.1 | 13 - 14 | 2406 | 58 | |
GenBank mRNA | AJ238540.1 | - | 13 - 14 | 58 | ||
AK145936.1 | - | 13 - 14 | 2387 | 58 | ||
BC054472.1 | - | 13 - 14 | 2325 | 6465367 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |