Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015732.4 | NP_056547.3 | 3 - 4 | 1300 | 67 | |
GenBank mRNA | AF073788.1 | - | 3 - 4 | 1165 | 67 | |
AF205889.1 | - | 2 - 3 | 1005 | 67 | ||
AK049192.1 | - | 3 - 4 | 1244 | 67 | ||
AK142333.1 | - | 3 - 4 | 1302 | 67 | ||
BC057338.1 | - | 3 - 4 | 1283 | 6827741 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |