Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293667.1 | NP_001280596.1 | 7 - 8 | 968 | 80 | |
NM_001293668.1 | NP_001280597.1 | 7 - 8 | 968 | 80 | ||
NM_015804.3 | NP_056619.1 | 7 - 8 | 968 | 80 | ||
NR_121603.1 | 7 - 8 | 968 | 80 | |||
GenBank mRNA | AF156551.1 | - | 7 - 8 | 760 | 80 | |
AK147497.1 | - | 7 - 8 | 971 | 80 | ||
AK173088.1 | - | 7 - 8 | 915 | 80 | ||
BC138715.1 | - | 7 - 8 | 687 | 8861737 | 80 | |
BC145399.1 | - | 7 - 8 | 688 | 80 | ||
BC145400.1 | - | 7 - 8 | 688 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |