Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015826.5 | NP_056641.2 | 2 - 3 | 685 | 117 | |
GenBank mRNA | AB221653.1 | - | 2 - 3 | 537 | 117 | |
AF192561.1 | - | 2 - 3 | 228 | 117 | ||
AF202778.1 | - | 2 - 3 | 672 | 117 | ||
AL133300.1 | - | 2 - 3 | 688 | 117 | ||
AY169786.1 | - | 2 - 3 | 688 | 117 | ||
AY169787.1 | - | 2 - 3 | 688 | 117 | ||
BC125474.1 | - | 2 - 3 | 648 | 40129989 | 117 | |
DQ530630.1 | - | 2 - 3 | 684 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |