Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114116.1 | NP_001107588.1 | 5 - 6 | 1263 | 104 | |
NM_016663.3 | NP_057872.3 | 5 - 6 | 1290 | 104 | ||
GenBank mRNA | AB000893.1 | - | 3 - 4 | 1053 | 104 | |
BC051969.1 | - | 4 - 5 | 1171 | 5694653 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |