Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114116.1 | NP_001107588.1 | 7 - 8 | 1603 | 68 | |
NM_016663.3 | NP_057872.3 | 7 - 8 | 1630 | 68 | ||
GenBank mRNA | AB000893.1 | - | 5 - 6 | 1393 | 68 | |
BC051969.1 | - | 6 - 7 | 1511 | 5694653 | 68 | |
D45858.1 | - | 6 - 7 | 1564 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |