Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00011141.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011141.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011141.mm.1 |
NM_016691.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011141.mm.1 |
NM_016691.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011141.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011144.mm.1 |
NM_016691.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011144.mm.1 |
NM_016691.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011144.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011154.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011154.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011154.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011147.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011147.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011147.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011153.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011153.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011153.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011146.mm.1 |
NM_016691.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011146.mm.1 |
NM_016691.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011146.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011152.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011152.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011152.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011140.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011140.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011140.mm.1 |
NM_016691.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011140.mm.1 |
NM_016691.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011140.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011149.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011149.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011149.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011142.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011142.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011142.mm.1 |
NM_016691.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011142.mm.1 |
NM_016691.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011142.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011143.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011143.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011143.mm.1 |
NM_016691.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011143.mm.1 |
NM_016691.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011143.mm.1 |
TC0X00001766.mm.1 |
MTA TC |