Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016846.3 | NP_058542.2 | 13 - 14 | 1571 | 106 | |
GenBank mRNA | AK144218.1 | - | 13 - 14 | 1646 | 106 | |
AK169109.1 | - | 13 - 14 | 1623 | 106 | ||
AK169180.1 | - | 13 - 14 | 1624 | 106 | ||
AK173071.1 | - | 3 - 4 | 166 | 106 | ||
BC017678.1 | - | 13 - 14 | 1571 | 4241244 | 106 | |
U14103.1 | - | 13 - 14 | 1629 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |