Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Corin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122756.1 | NP_001116228.1 | 17 - 18 | 2419 | 79 | |
NM_016869.3 | NP_058565.2 | 17 - 18 | 2576 | 79 | ||
GenBank mRNA | AB013874.1 | - | 17 - 18 | 2576 | 79 | |
BC093485.1 | - | 17 - 18 | 2352 | 30648458 | 79 | |
BC138339.1 | - | 17 - 18 | 2559 | 8861359 | 79 | |
BC138340.1 | - | 17 - 18 | 2559 | 8861360 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |