Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31925370 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31925369 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31925368 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P400224 |
CF929049.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P400224 |
AF173379.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P400224 |
AK010264.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P400224 |
AK050072.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P400224 |
NM_016892.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P400224 |
CF929049.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P400224 |
AF173379.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P400224 |
AK010264.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P400224 |
AK050072.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P400224 |
NM_016892.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
718661 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
382545 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347367 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770678 |
NM_001105181.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770678 |
XM_885089.3 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770678 |
XM_913169.3 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
443950 |
AK046640.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
536119 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
733923 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
154800 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
38225 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P153725 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P117415 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P400224 |
CF929049.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P400224 |
AF173379.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P400224 |
AK010264.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P400224 |
AK050072.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P400224 |
NM_016892.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P400224 |
CF929049.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P400224 |
AF173379.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P400224 |
AK010264.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P400224 |
AK050072.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P400224 |
NM_016892.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P193536 |
AF173379.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P193536 |
AK050072.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P193536 |
NM_016892.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P193536 |
AF121906.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P193536 |
BC026938.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P400224 |
CF929049.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P400224 |
AF173379.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P400224 |
AK010264.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P400224 |
AK050072.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P400224 |
NM_016892.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P400224 |
CF929049.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P400224 |
AF173379.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P400224 |
AK010264.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P400224 |
AK050072.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P400224 |
NM_016892.3 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925350 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925345 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925353 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925346 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925366 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925344 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925361 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925356 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925359 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925362 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925354 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925351 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925369 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925365 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925348 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925367 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925363 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925343 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925368 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31925347 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P153725 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03541915 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P193536 |
AF173379.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P193536 |
AK050072.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P193536 |
NM_016892.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P193536 |
AF121906.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P193536 |
BC026938.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P400224 |
CF929049.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P400224 |
AF173379.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P400224 |
AK010264.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P400224 |
AK050072.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P400224 |
NM_016892.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P400224 |
CF929049.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P400224 |
AF173379.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P400224 |
AK010264.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P400224 |
AK050072.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P400224 |
NM_016892.3 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136239 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P127087 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P153725 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14186167 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14186185 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14186202 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14186176 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14186205 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31925368 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31925370 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31925369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925325 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925321 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925341 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925339 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925345 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925292 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925328 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925368 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925295 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925333 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925306 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925315 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925299 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925314 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925313 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925303 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925330 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925365 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925290 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925291 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925343 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925298 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925329 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31925296 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925366 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925332 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14182849 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925369 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925359 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925361 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925330 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925365 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14148744 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925343 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925346 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925367 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14372275 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14182850 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925368 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925354 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925356 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925363 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925349 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14340846 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14148749 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925336 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925350 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925355 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925351 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925345 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925328 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31925339 |
- |
- |