Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243049.1 | NP_001229978.1 | 9 - 10 | 1020 | 86 | |
NM_001243050.1 | NP_001229979.1 | 10 - 11 | 1198 | 86 | ||
NM_001243051.1 | NP_001229980.1 | 10 - 11 | 1219 | 86 | ||
NM_016920.3 | NP_058616.1 | 10 - 11 | 1198 | 86 | ||
GenBank mRNA | AB022321.1 | - | 9 - 10 | 1027 | 86 | |
AF218249.1 | - | 10 - 11 | 1168 | 86 | ||
AF218250.1 | - | 9 - 10 | 1042 | 86 | ||
AF218251.1 | - | 9 - 10 | 1021 | 86 | ||
AK029316.1 | - | 9 - 10 | 2952 | 86 | ||
AK158786.1 | - | 9 - 10 | 988 | 86 | ||
AK159114.1 | - | 10 - 11 | 1134 | 86 | ||
AK172078.1 | - | 10 - 11 | 1128 | 86 | ||
BC066839.1 | - | 10 - 11 | 1163 | 30101340 | 86 | |
BC071182.1 | - | 10 - 11 | 1163 | 30101184 | 86 | |
U13836.1 | - | 9 - 10 | 1060 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |