Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100003998.mm.1 |
NM_001163671.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003998.mm.1 |
NM_001163671.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003998.mm.1 |
NM_001163672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003998.mm.1 |
NM_001163672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003998.mm.1 |
NM_016961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003998.mm.1 |
NM_016961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003998.mm.1 |
NM_207692.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003998.mm.1 |
NM_207692.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003998.mm.1 |
TC1100000527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004002.mm.1 |
NM_001163671.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004002.mm.1 |
NM_001163671.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004002.mm.1 |
NM_001163672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004002.mm.1 |
NM_001163672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004002.mm.1 |
NM_016961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004002.mm.1 |
NM_016961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004002.mm.1 |
NM_207692.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004002.mm.1 |
NM_207692.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004002.mm.1 |
TC1100000527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100003999.mm.1 |
NM_001163671.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003999.mm.1 |
NM_001163671.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003999.mm.1 |
NM_001163672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003999.mm.1 |
NM_001163672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003999.mm.1 |
NM_016961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003999.mm.1 |
NM_016961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003999.mm.1 |
NM_207692.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003999.mm.1 |
NM_207692.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003999.mm.1 |
TC1100000527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004001.mm.1 |
NM_001163671.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004001.mm.1 |
NM_001163671.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004001.mm.1 |
NM_001163672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004001.mm.1 |
NM_001163672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004001.mm.1 |
NM_016961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004001.mm.1 |
NM_016961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004001.mm.1 |
NM_207692.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004001.mm.1 |
NM_207692.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004001.mm.1 |
TC1100000527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004004.mm.1 |
NM_001163671.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004004.mm.1 |
NM_001163671.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004004.mm.1 |
NM_001163672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004004.mm.1 |
NM_001163672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004004.mm.1 |
NM_016961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004004.mm.1 |
NM_016961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004004.mm.1 |
NM_207692.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004004.mm.1 |
NM_207692.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004004.mm.1 |
TC1100000527.mm.1 |
MTA TC |