Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290627.1 | NP_001277556.1 | 9 - 10 | 1407 | 84 | |
NM_018731.3 | NP_061201.2 | 9 - 10 | 1407 | 84 | ||
GenBank mRNA | AK143905.1 | - | 9 - 10 | 1408 | 84 | |
BC015262.1 | - | 9 - 10 | 1393 | 4241977 | 84 | |
U17282.1 | - | 8 - 9 | 1374 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |