Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB026804.1 | - | 4 - 5 | 353 | 64 | |
AB075900.1 | - | 5 - 6 | 485 | 64 | ||
AB075901.1 | - | 7 - 8 | 698 | 64 | ||
BC105660.1 | - | 4 - 5 | 352 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |