Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019445.2 | NP_062318.2 | 13 - 14 | 4550 | 106 | |
GenBank mRNA | AF218940.1 | - | 14 - 15 | 4512 | 106 | |
AK140268.1 | - | 7 - 8 | 835 | 106 | ||
BC094606.1 | - | 13 - 14 | 4550 | 6840339 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |