Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291071.1 | NP_001278000.1 | 20 - 21 | 2211 | 72 | |
NM_019703.4 | NP_062677.1 | 20 - 21 | 2211 | 72 | ||
GenBank mRNA | AF123533.1 | - | 20 - 21 | 2176 | 72 | |
AK004790.1 | - | 20 - 21 | 2165 | 72 | ||
AK047303.1 | - | 2 - 3 | 210 | 72 | ||
AK049841.1 | - | 20 - 21 | 2165 | 72 | ||
AK076803.1 | - | 20 - 21 | 2166 | 72 | ||
AK098162.1 | - | 20 - 21 | 2165 | 72 | ||
AK152670.1 | - | 20 - 21 | 2166 | 72 | ||
AK170624.1 | - | 20 - 21 | 2166 | 72 | ||
AK171062.1 | - | 20 - 21 | 2166 | 72 | ||
AY779275.1 | - | 20 - 21 | 2128 | 72 | ||
AY779276.1 | - | 20 - 21 | 2128 | 72 | ||
BC006926.1 | - | 20 - 21 | 2150 | 2655383 | 72 | |
CT010268.1 | - | 20 - 21 | 2119 | 72 | ||
Y19008.1 | - | 20 - 21 | 2168 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |