Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019727.2 | NP_062701.2 | 8 - 9 | 928 | 76 | |
GenBank mRNA | AB019214.1 | - | 8 - 9 | 868 | 76 | |
AF154120.1 | - | 8 - 9 | 932 | 76 | ||
AK149850.1 | - | 8 - 9 | 840 | 76 | ||
AK154079.1 | - | 8 - 9 | 841 | 76 | ||
AK155746.1 | - | 8 - 9 | 816 | 76 | ||
AK161241.1 | - | 8 - 9 | 909 | 76 | ||
AK167177.1 | - | 8 - 9 | 883 | 76 | ||
AK167993.1 | - | 8 - 9 | 823 | 76 | ||
AK168742.1 | - | 8 - 9 | 921 | 76 | ||
AK192993.1 | - | 2 - 3 | 118 | 76 | ||
BC066044.1 | - | 8 - 9 | 885 | 6807363 | 76 | |
BC066189.1 | - | 8 - 9 | 819 | 30471747 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |