Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500027366.mm.1 |
TC0500003066.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500027366.mm.1 |
NM_001077359.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027366.mm.1 |
NM_001077359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027366.mm.1 |
NM_001077360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027366.mm.1 |
NM_001077360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027366.mm.1 |
NM_019834.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027366.mm.1 |
NM_019834.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027367.mm.1 |
TC0500003066.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500027367.mm.1 |
NM_001077359.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027367.mm.1 |
NM_001077359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027367.mm.1 |
NM_001077360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027367.mm.1 |
NM_001077360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027367.mm.1 |
NM_019834.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027367.mm.1 |
NM_019834.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027369.mm.1 |
TC0500003066.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500027369.mm.1 |
NM_001077359.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027369.mm.1 |
NM_001077359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027369.mm.1 |
NM_001077360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027369.mm.1 |
NM_001077360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027369.mm.1 |
NM_019834.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027369.mm.1 |
NM_019834.3 |
Refseq |