Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cftr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021050.2 | NP_066388.1 | 13 - 14 | 1899 | 149 | |
GenBank mRNA | M60493.1 | - | 13 - 14 | 1900 | 149 | |
M69298.1 | - | 13 - 14 | 1898 | 149 | ||
X72694.1 | - | 3 - 4 | 227 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mus musculus Probe spans exons |