Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122733.1 | NP_001116205.1 | 1 - 2 | 136 | 104 | |
NM_021099.3 | NP_066922.2 | 1 - 2 | 132 | 104 | ||
GenBank mRNA | AK046795.1 | - | 1 - 2 | 128 | 104 | |
AK047010.1 | - | 1 - 2 | 130 | 104 | ||
AY536430.1 | - | 1 - 2 | 95 | 104 | ||
AY536431.1 | - | 1 - 2 | 95 | 104 | ||
BC026713.1 | - | 1 - 2 | 115 | 104 | ||
BC052457.1 | - | 1 - 2 | 107 | 6830637 | 104 | |
BC075716.1 | - | 1 - 2 | 121 | 3673641 | 104 | |
BY123520.1 | - | 1 - 2 | 136 | 104 | ||
Y00864.1 | - | 1 - 2 | 99 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |