Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021305.3 | NP_067280.1 | 7 - 8 | 713 | 93 | |
GenBank mRNA | AF145254.1 | - | 7 - 8 | 649 | 93 | |
AF222748.1 | - | 7 - 8 | 650 | 93 | ||
AK017615.1 | - | 7 - 8 | 695 | 93 | ||
AK160602.1 | - | 7 - 8 | 697 | 93 | ||
BC005458.1 | - | 7 - 8 | 656 | 3494001 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |