Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700015156.mm.1 |
NM_001163691.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015156.mm.1 |
NM_001163691.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015156.mm.1 |
NM_021415.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015156.mm.1 |
NM_021415.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015156.mm.1 |
TC1700001661.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700015160.mm.1 |
NM_001163691.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015160.mm.1 |
NM_001163691.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015160.mm.1 |
NM_021415.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015160.mm.1 |
NM_021415.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015160.mm.1 |
TC1700001661.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700015161.mm.1 |
NM_001163691.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015161.mm.1 |
NM_001163691.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015161.mm.1 |
NM_021415.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015161.mm.1 |
NM_021415.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015161.mm.1 |
TC1700001661.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700015158.mm.1 |
NM_001163691.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015158.mm.1 |
NM_001163691.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015158.mm.1 |
NM_021415.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015158.mm.1 |
NM_021415.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015158.mm.1 |
TC1700001661.mm.1 |
MTA TC |