Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163691.1 | NP_001157163.1 | 23 - 24 | 4840 | 67 | |
NM_021415.4 | NP_067390.4 | 23 - 24 | 4840 | 67 | ||
GenBank mRNA | AK122452.1 | - | 12 - 13 | 1813 | 67 | |
AK135951.1 | - | 5 - 6 | 587 | 67 | ||
AK147242.1 | - | 23 - 24 | 4841 | 67 | ||
BC138026.1 | - | 23 - 24 | 4679 | 8861045 | 67 | |
BU612176.1 | - | 1 - 2 | 66 | |||
CB246265.1 | - | 2 - 3 | 253 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |