Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cse1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023565.3 | NP_076054.1 | 19 - 20 | 2275 | 79 | |
GenBank mRNA | AF301152.1 | - | 19 - 20 | 2275 | 79 | |
AK138940.1 | - | 5 - 6 | 699 | 79 | ||
BC145393.1 | - | 18 - 19 | 2046 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |