Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025711.3 | NP_079987.2 | 1 - 2 | 274 | 93 | |
GenBank mRNA | AK014504.1 | - | 1 - 2 | 230 | 93 | |
AK021386.1 | - | 1 - 2 | 229 | 93 | ||
AK085920.1 | - | 1 - 2 | 229 | 93 | ||
AW106088.1 | - | 1 - 2 | 274 | 93 | ||
BC034888.1 | - | 1 - 2 | 176 | 1365428 | 93 | |
BC145904.1 | - | 1 - 2 | 172 | 40131207 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |