Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199271.1 | NP_001186200.1 | 4 - 5 | 1221 | 75 | |
NM_031196.3 | NP_112473.1 | 4 - 5 | 1315 | 75 | ||
GenBank mRNA | AK002560.1 | - | 4 - 5 | 1232 | 75 | |
AK079592.1 | - | 4 - 5 | 1276 | 75 | ||
AK085024.2 | - | 4 - 5 | 1263 | 75 | ||
AK088803.1 | - | 4 - 5 | 1278 | 75 | ||
AK138577.1 | - | 4 - 5 | 1141 | 75 | ||
BC015263.1 | - | 4 - 5 | 1233 | 4239690 | 75 | |
L23755.1 | - | 3 - 4 | 1173 | 75 | ||
L36539.1 | - | 4 - 5 | 1228 | 75 | ||
U32469.1 | - | 4 - 5 | 1255 | 75 | ||
U66103.1 | - | 4 - 5 | 1268 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |