Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031197.2 | NP_112474.2 | 7 - 8 | 1072 | 92 | |
GenBank mRNA | AK005068.1 | - | 7 - 8 | 1073 | 92 | |
AK165430.1 | - | 7 - 8 | 1007 | 92 | ||
BC034675.1 | - | 7 - 8 | 991 | 4236331 | 92 | |
X15684.1 | - | 7 - 8 | 1021 | 92 | ||
X16986.1 | - | 7 - 8 | 1011 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |