Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033444.2 | NP_254279.1 | 2 - 3 | 356 | 101 | |
GenBank mRNA | AK088809.1 | - | 2 - 3 | 167 | 101 | |
AK150382.1 | - | 2 - 3 | 634 | 101 | ||
AK150611.1 | - | 2 - 3 | 345 | 101 | ||
AK153509.1 | - | 2 - 3 | 634 | 101 | ||
AK167765.1 | - | 2 - 3 | 362 | 101 | ||
AK168262.1 | - | 2 - 3 | 176 | 101 | ||
BC004658.1 | - | 2 - 3 | 356 | 3498118 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |