Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080465.2 | NP_536713.1 | 1 - 2 | 475 | 101 | |
GenBank mRNA | AF357240.1 | - | 1 - 2 | 475 | 101 | |
AF533008.1 | - | 1 - 2 | 475 | 101 | ||
AK033158.1 | - | 4 - 5 | 1979 | 101 | ||
AK039033.1 | - | 1 - 2 | 1161 | 101 | ||
AK050390.1 | - | 1 - 2 | 731 | 101 | ||
AK087908.1 | - | 1 - 2 | 820 | 101 | ||
AK149082.1 | - | 1 - 2 | 1163 | 101 | ||
AY123778.1 | - | 1 - 2 | 550 | 101 | ||
AY280964.1 | - | 1 - 2 | 475 | 101 | ||
BC125475.1 | - | 1 - 2 | 501 | 40129990 | 101 | |
BC137656.1 | - | 1 - 2 | 501 | 8860675 | 101 | |
EU478850.1 | - | 1 - 2 | 475 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |