Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289564.1 | NP_001276493.1 | 9 - 10 | 926 | 79 | |
NM_001289566.1 | NP_001276495.1 | 9 - 10 | 968 | 79 | ||
NM_080468.2 | NP_536716.2 | 9 - 10 | 926 | 79 | ||
GenBank mRNA | AF346501.1 | - | 9 - 10 | 926 | 79 | |
AK034592.1 | - | 9 - 10 | 839 | 79 | ||
BC120741.1 | - | 9 - 10 | 895 | 40129664 | 79 | |
BC144802.1 | - | 9 - 10 | 895 | 9053350 | 79 | |
BC144803.1 | - | 9 - 10 | 937 | 9053351 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |