Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138650.2 | NP_619591.1 | 13 - 14 | 1654 | 67 | |
GenBank mRNA | AK083258.1 | - | 12 - 13 | 1530 | 67 | |
AK087972.1 | - | 13 - 14 | 1865 | 67 | ||
AK215466.1 | - | 1 - 2 | 91 | 67 | ||
AK220294.1 | - | 13 - 14 | 1596 | 67 | ||
BC015278.1 | - | 12 - 13 | 1227 | 4221290 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |