Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144520.2 | NP_653103.1 | 3 - 4 | 224 | 76 | |
GenBank mRNA | BC005759.1 | - | 3 - 4 | 224 | 2655022 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |