Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004701.2 | - | 8 - 9 | 1084 | 108 | |
AK144750.1 | - | 9 - 10 | 1041 | 108 | ||
BC003816.1 | - | 9 - 10 | 1081 | 3485483 | 108 | |
M72394.1 | - | 9 - 10 | 1029 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |