Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcea2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009326.2 | NP_033352.1 | 7 - 8 | 772 | 76 | |
GenBank mRNA | AB010271.1 | - | 7 - 8 | 710 | 76 | |
AB221557.1 | - | 7 - 8 | 675 | 76 | ||
AK131650.1 | - | 5 - 6 | 1647 | 76 | ||
AK137527.1 | - | 7 - 8 | 654 | 76 | ||
BC049617.1 | - | 7 - 8 | 765 | 6773546 | 76 | |
BC065786.1 | - | 7 - 8 | 763 | 5120316 | 76 | |
BC138504.1 | - | 7 - 8 | 761 | 8861524 | 76 | |
BC138505.1 | - | 7 - 8 | 761 | 8861525 | 76 | |
D86081.1 | - | 7 - 8 | 772 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |