Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Th | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009377.1 | NP_033403.1 | 5 - 6 | 655 | 81 | |
GenBank mRNA | AK139568.1 | - | 872 | 81 | ||
M69200.1 | - | 5 - 6 | 655 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |