Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009484.2 | NP_033510.2 | 1 - 2 | 334 | 64 | |
GenBank mRNA | AF057367.1 | - | 1 - 2 | 314 | 64 | |
AK043154.1 | - | 1 - 2 | 334 | 64 | ||
AK142344.1 | - | 2 - 3 | 266 | 64 | ||
AK155922.1 | - | 2 - 3 | 214 | 64 | ||
AK162410.1 | - | 2 - 3 | 201 | 64 | ||
AK220463.1 | - | 1 - 2 | 290 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |