Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009484.2 | NP_033510.2 | 2 - 3 | 394 | 90 | |
GenBank mRNA | AF057367.1 | - | 2 - 3 | 374 | 90 | |
AK043154.1 | - | 2 - 3 | 394 | 90 | ||
AK142512.1 | - | 2 - 3 | 310 | 90 | ||
AK155922.1 | - | 3 - 4 | 274 | 90 | ||
AK220463.1 | - | 2 - 3 | 350 | 90 | ||
BC053061.1 | - | 2 - 3 | 377 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |