Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009484.2 | NP_033510.2 | 13 - 14 | 1504 | 68 | |
GenBank mRNA | AF057367.1 | - | 13 - 14 | 1484 | 68 | |
AK142344.1 | - | 15 - 16 | 1588 | 68 | ||
AK142512.1 | - | 13 - 14 | 1420 | 68 | ||
BC053061.1 | - | 13 - 14 | 1487 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |